This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01332260_m1
See other PAK3 GE Assays ›
Gene Symbol:
Pak3
Gene Name:
p21 protein (Cdc42/Rac)-activated kinase 3
Gene Aliases:
PAK-3, Pak65alpha, Pak65beta, Stk4
Chromosome Location:
Chr.X: 143518368 - 143797796 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01332260_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pak3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195046.1 NP_001181975.1 1 - 2 205 77
NM_001195047.1 NP_001181976.1 1 - 2 205 77
NM_001195048.1 NP_001181977.1 2 - 3 378 77
NM_001195049.1 NP_001181978.1 5 - 6 687 77
NM_008778.3 NP_032804.2 2 - 3 296 77
GenBank mRNA AF082297.1 - 1 - 2 210 77
AJ496262.1 - 1 - 2 178 77
AJ496263.1 - 1 - 2 178 77
AK031853.1 - 2 - 3 259 77
AK158893.1 - 2 - 3 379 77
AM943853.1 - 1 - 2 178 77
AM943854.1 - 1 - 2 178 77
BC053403.1 - 2 - 3 303 30060082 77
U39738.1 - 5 - 6 694 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00001413.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pak3

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
activation of MAPK activity
protein phosphorylation
Rho protein signal transduction
multicellular organism development
regulation of mitotic cell cycle
axonogenesis
metabolic process
regulation of neuron projection development
phosphorylation
dendrite development
cell migration
signal transduction by protein phosphorylation
actin cytoskeleton organization
regulation of actin filament polymerization
stress-activated protein kinase signaling cascade
activation of protein kinase activity
regulation of apoptotic process
regulation of MAPK cascade
positive regulation of neuron apoptotic process
dendritic spine development
positive regulation of dendritic spine morphogenesis
cellular response to organic cyclic compound
positive regulation of DNA biosynthetic process
nucleotide binding
catalytic activity
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
Rho GTPase binding
SH3 domain binding
metal ion binding
Rac GTPase binding
GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00008961.mm.1 NM_001195046.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008961.mm.1 NM_001195046.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008961.mm.1 NM_001195047.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008961.mm.1 NM_001195047.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008961.mm.1 NM_001195048.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008961.mm.1 NM_001195048.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008961.mm.1 NM_001195049.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008961.mm.1 NM_001195049.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008961.mm.1 NM_008778.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008961.mm.1 NM_008778.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008961.mm.1 TC0X00001413.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00008938.mm.1 NM_001195048.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008938.mm.1 NM_001195048.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008938.mm.1 TC0X00001413.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00008963.mm.1 NM_001195046.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008963.mm.1 NM_001195046.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008963.mm.1 NM_001195048.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008963.mm.1 NM_001195048.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008963.mm.1 TC0X00001413.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00008939.mm.1 NM_001195048.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008939.mm.1 NM_001195048.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008939.mm.1 TC0X00001413.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00008962.mm.1 NM_001195046.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008962.mm.1 NM_001195046.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008962.mm.1 NM_001195047.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008962.mm.1 NM_001195047.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008962.mm.1 TC0X00001413.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00008942.mm.1 NM_008778.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008942.mm.1 TC0X00001413.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0