This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01332314_g1
See other LAIR1 GE Assays ›
Gene Symbol:
Lair1
Gene Name:
leukocyte-associated Ig-like receptor 1
Gene Aliases:
5133400O11Rik, BB115266, D7Bwg0421e, Lair-1
Chromosome Location:
Chr.7: 4007073 - 4073507 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01332314_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lair1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113474.1 NP_001106945.1 5 - 6 658 77
NM_178611.6 NP_848726.1 4 - 5 328 77
GenBank mRNA AF479685.1 - 5 - 6 593 77
AF491829.1 - 5 - 6 691 77
AF491830.1 - 4 - 5 240 77
AF491831.1 - 5 - 6 570 77
AK033472.1 - 4 - 5 305 77
AK038574.1 - 4 - 5 291 77
AK043664.1 - 4 - 5 385 77
AK044465.1 - 5 - 6 629 77
AK050281.1 - 5 - 6 614 77
AK054158.1 - 5 - 6 737 77
AY392763.1 - 5 - 6 595 77
AY392764.1 - 4 - 5 265 77
AY392765.1 - 3 - 4 148 77
BC025901.1 - 5 - 6 622 5253846 77
BY204263.1 - 3 - 4 272 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lair1

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0