This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01332698_g1
See other CKB GE Assays ›
Gene Symbol:
Ckb
Gene Name:
creatine kinase, brain
Gene Aliases:
B-CK, Bck, Ck-3, Ck3, Ckbb
Chromosome Location:
Chr.12: 111669355 - 111672338 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01332698_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ckb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021273.4 NP_067248.1 4 - 5 600 105
GenBank mRNA AK002467.1 - 4 - 5 563 105
AK014299.1 - 4 - 5 543 105
AK148885.1 - 4 - 5 560 105
AK152388.1 - 4 - 5 560 105
AK153484.1 - 4 - 5 560 105
AK161990.1 - 4 - 5 560 105
AK166980.1 - 4 - 5 563 105
AK167034.1 - 4 - 5 563 105
BC015271.1 - 4 - 5 530 4225384 105
BC106109.1 - 4 - 5 527 6395794 105

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ckb

Panther Classification:

Molecular Function -

amino acid kinase

Gene Ontology Categories:

Function(s) Process(es)

phosphorylation
cellular chloride ion homeostasis
nucleotide binding
catalytic activity
creatine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
ubiquitin protein ligase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200016733.mm.1 NM_021273.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200016733.mm.1 NM_021273.4 Refseq
Affymetrix Mouse Clariom S Array PSR1200016733.mm.1 TC1200002411.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200016735.mm.1 NM_021273.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200016735.mm.1 NM_021273.4 Refseq
Affymetrix Mouse Clariom S Array PSR1200016735.mm.1 TC1200002411.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-wtj7v:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline