This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01333193_g1
See other UCMA GE Assays ›
Gene Symbol:
Ucma
Gene Name:
upper zone of growth plate and cartilage matrix associated
Gene Aliases:
1110017I16Rik, AW121955, Grp
Chromosome Location:
Chr.2: 4976122 - 4985748 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01333193_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ucma
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113558.2 NP_001107030.1 1 - 2 194 83
NM_001165932.1 NP_001159404.1 1 - 2 194 83
GenBank mRNA BC115608.2 - 1 - 2 97 40102702 83
BC115609.2 - 1 - 2 97 40102703 83
BE648935.1 - 1 - 2 157 83
EF529510.1 - 1 - 2 165 83
EU368184.1 - 1 - 2 194 83
FJ217397.1 - 1 - 2 132 83
FJ217399.1 - 1 - 2 132 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000040.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of osteoblast differentiation
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200000319.mm.1 NM_001113558.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000319.mm.1 NM_001113558.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200000319.mm.1 NM_001165932.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000319.mm.1 NM_001165932.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000319.mm.1 TC0200000040.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0