This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01333271_m1
See other DCLRE1C GE Assays ›
Gene Symbol:
Dclre1c
Gene Name:
DNA cross-link repair 1C, PSO2 homolog (S. cerevisiae)
Gene Aliases:
9930121L06Rik, AI661365, Art, Snm1l
Chromosome Location:
Chr.2: 3424062 - 3474330 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01333271_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dclre1c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110214.1 NP_001103684.1 14 - 15 1280 152
GenBank mRNA BC108935.2 - 14 - 15 1233 40039591 152

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000006.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

telomere maintenance
immune system process
DNA metabolic process
DNA repair
double-strand break repair
DNA recombination
cellular response to DNA damage stimulus
response to ionizing radiation
B cell differentiation
V(D)J recombination
chromosome organization
nucleic acid phosphodiester bond hydrolysis
single-stranded DNA endodeoxyribonuclease activity
nuclease activity
endonuclease activity
exonuclease activity
5'-3' exonuclease activity
hydrolase activity
phosphoric ester hydrolase activity
T/G mismatch-specific endonuclease activity
retroviral 3' processing activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200000093.mm.1 NM_001110214.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000093.mm.1 NM_001110214.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000093.mm.1 NM_175683.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000093.mm.1 NM_175683.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200000093.mm.1 TC0200000006.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200000092.mm.1 NM_001110214.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000092.mm.1 NM_001110214.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000092.mm.1 NM_175683.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000092.mm.1 NM_175683.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200000092.mm.1 TC0200000006.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200000091.mm.1 NM_001110214.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000091.mm.1 NM_001110214.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000091.mm.1 NM_175683.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000091.mm.1 NM_175683.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200000091.mm.1 TC0200000006.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200000090.mm.1 NM_001110214.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000090.mm.1 NM_001110214.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000090.mm.1 NM_175683.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000090.mm.1 NM_175683.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200000090.mm.1 TC0200000006.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200000089.mm.1 NM_001110214.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000089.mm.1 NM_001110214.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000089.mm.1 NM_175683.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000089.mm.1 NM_175683.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200000089.mm.1 TC0200000006.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 591038 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0