This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01333274_m1
See other NUDT5 GE Assays ›
Gene Symbol:
Nudt5
Gene Name:
nudix (nucleoside diphosphate linked moiety X)-type motif 5
Gene Aliases:
-
Chromosome Location:
Chr.2: 5845034 - 5868745 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01333274_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nudt5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016918.3 NP_058614.1 3 - 4 521 112
GenBank mRNA AF222786.1 - 2 - 3 171 112
AK088222.1 - 3 - 4 522 112
BC004571.1 - 3 - 4 262 3586340 112
BC049595.1 - 3 - 4 326 6774692 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000055.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nudt5

Panther Classification:

Molecular Function -

pyrophosphatase

Gene Ontology Categories:

Function(s) Process(es)

nucleobase-containing compound metabolic process
ribonucleoside diphosphate catabolic process
magnesium ion binding
RNA binding
hydrolase activity
nucleoside-diphosphatase activity
ADP-sugar diphosphatase activity
snoRNA binding
metal ion binding
ADP-ribose diphosphatase activity
m7G(5')pppN diphosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200000359.mm.1 NM_016918.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000359.mm.1 NM_016918.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200000359.mm.1 TC0200000055.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0