This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01333388_m1
See other IL15RA GE Assays ›
Gene Symbol:
Il15ra
Gene Name:
interleukin 15 receptor, alpha chain
Gene Aliases:
AA690181, IL-15RA
Chromosome Location:
Chr.2: 11705169 - 11733985 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01333388_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il15ra
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271499.1 NP_001258428.1 2 - 3 430 81
NM_001271500.1 NP_001258429.1 2 - 3 430 81
NM_008358.2 NP_032384.1 2 - 3 430 81
NM_133836.2 NP_598597.1 2 - 3 483 81
NR_073191.1 2 - 3 483 81
GenBank mRNA AY219715.1 - 2 - 3 331 81
AY219716.1 - 2 - 3 332 81
AY221617.1 - 2 - 3 224 81
BC022705.1 - 2 - 3 240 81
BC069881.1 - 2 - 3 389 81

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AY221617.1, BC022705.1, NM_133836.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Il15ra

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s)

JAK-STAT cascade
negative regulation of neuron projection development
positive regulation of natural killer cell differentiation

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l2rv2:80/100.66.78.26:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0