Assay Details
| Gene Symbol: Il15ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271500.1 | NP_001258429.1 | 3 - 4 | 521 | 97 | |
| NR_073191.1 | 3 - 4 | 574 | 97 | |||
| GenBank mRNA | AY219716.1 | - | 3 - 4 | 423 | 97 | |
| AY221617.1 | - | 3 - 4 | 315 | 97 | ||
| BC069881.1 | - | 3 - 4 | 480 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY221617.1 transcripts.
Set Membership: |
|

Il15ra
Genomic Map