This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01333420_mH
See other STAM GE Assays ›
Gene Symbol:
Stam
Gene Name:
signal transducing adaptor molecule (SH3 domain and ITAM motif) 1
Gene Aliases:
STAM1
Chromosome Location:
Chr.2: 14073646 - 14149210 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01333420_mH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stam
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011484.2 NP_035614.1 5 - 6 628 64
GenBank mRNA AK035924.1 - 5 - 6 551 64
AK147832.1 - 5 - 6 538 64
AK157767.1 - 5 - 6 575 64
AK163720.1 - 5 - 6 575 64
AK163922.1 - 5 - 6 565 64
AK169170.1 - 5 - 6 635 64
BC044666.1 - 5 - 6 628 4950569 64
BC055326.1 - 5 - 6 574 6416552 64
U43900.1 - 5 - 6 485 64

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stam

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
protein transport
positive regulation of exosomal secretion
regulation of extracellular exosome assembly
protein binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 300862 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline