This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01333421_mH
See other STAM GE Assays ›
Gene Symbol:
Stam
Gene Name:
signal transducing adaptor molecule (SH3 domain and ITAM motif) 1
Gene Aliases:
STAM1
Chromosome Location:
Chr.2: 14073646 - 14149210 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01333421_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stam
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011484.2 NP_035614.1 8 - 9 1003 117
GenBank mRNA AK035924.1 - 8 - 9 926 117
AK144987.1 - 8 - 9 1020 117
AK147832.1 - 8 - 9 913 117
AK157767.1 - 8 - 9 950 117
AK162431.1 - 8 - 9 950 117
AK163720.1 - 8 - 9 953 119
AK163922.1 - 8 - 9 940 117
AK169170.1 - 8 - 9 1010 117
BC044666.1 - 8 - 9 1003 4950569 117
BC055326.1 - 8 - 9 949 6416552 117
U43900.1 - 8 - 9 860 117

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stam

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
protein transport
positive regulation of exosomal secretion
regulation of extracellular exosome assembly
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200001000.mm.1 NM_011484.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200001000.mm.1 NM_011484.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200001000.mm.1 TC0200000242.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200001005.mm.1 NM_011484.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200001005.mm.1 NM_011484.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200001005.mm.1 TC0200000242.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline