This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01334780_m1
See other PPP1R1C GE Assays ›
Gene Symbol:
Ppp1r1c
Gene Name:
protein phosphatase 1, regulatory (inhibitor) subunit 1C
Gene Aliases:
4930565M23Rik, 4930579P15Rik
Chromosome Location:
Chr.2: 79707780 - 79818546 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01334780_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r1c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290743.1 NP_001277672.1 3 - 4 573 111
NM_172420.4 NP_766008.2 3 - 4 573 111
NR_110979.1 3 - 4 568 111
GenBank mRNA AK051670.1 - 3 - 4 574 111
AK083940.1 - 3 - 4 287 111
AK157871.1 - 3 - 4 432 111
BC120706.1 - 3 - 4 535 40129629 111
BC132149.1 - 3 - 4 535 40130426 111

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001166.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp1r1c

Panther Classification:

Molecular Function -

phosphatase inhibitor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein kinase activity
cell cycle
signal transduction
positive regulation of cell growth
intracellular signal transduction
negative regulation of catalytic activity
cell division
positive regulation of G1/S transition of mitotic cell cycle
protein phosphatase inhibitor activity
protein serine/threonine phosphatase inhibitor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200009376.mm.1 NM_172420.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200009376.mm.1 TC0200001166.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0