This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01335094_g1
See other GPAT2 GE Assays ›
Gene Symbol:
Gpat2
Gene Name:
glycerol-3-phosphate acyltransferase 2, mitochondrial
Gene Aliases:
A530057A03Rik, Gm116, xGPAT1
Chromosome Location:
Chr.2: 127410628 - 127436093 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01335094_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpat2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081089.2 NP_001074558.2 16 - 17 1931 69
GenBank mRNA AB257502.1 - 15 - 16 1849 69
AK040991.1 - 2 - 3 397 69
AK131154.1 - 16 - 17 1930 69
BC113165.1 - 16 - 17 1925 40059976 69
BC113776.1 - 16 - 17 1922 40059969 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001916.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycerol-3-phosphate metabolic process
lipid metabolic process
metabolic process
phospholipid biosynthetic process
triglyceride biosynthetic process
cellular lipid metabolic process
glycerol-3-phosphate O-acyltransferase activity
O-acyltransferase activity
transferase activity
transferase activity, transferring acyl groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200015040.mm.1 NM_001081089.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200015040.mm.1 TC0200001916.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200015039.mm.1 NM_001081089.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200015039.mm.1 TC0200001916.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0