This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01335342_g1
See other XIRP2 GE Assays ›
Gene Symbol:
Xirp2
Gene Name:
xin actin-binding repeat containing 2
Gene Aliases:
2310003D02Rik, 2310008C07Rik, A530024P18Rik, AI452089, Cmya3, Gm352, Xin2
Chromosome Location:
Chr.2: 67446000 - 67526620 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01335342_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Xirp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001024618.2 NP_001019789.1 4 - 5 532 124
NM_001083919.1 NP_001077388.1 4 - 5 532 124
GenBank mRNA AK040786.1 - 4 - 5 533 124
AY775570.1 - 3 - 4 386 124
DQ011666.1 - 4 - 5 533 124
EF119712.1 - 4 - 5 532 124
EF119713.1 - 688 124
EF119714.1 - 736 124
EF119715.1 - 763 124
EF119716.1 - 4 - 5 532 124
EF122140.1 - 3 - 4 287 124
KM273012.1 - 974 124

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000966.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Xirp2

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

ventricular septum development
heart development
actin cytoskeleton organization
cell-cell junction organization
cardiac muscle tissue morphogenesis
actin binding
protein binding
zinc ion binding
metal ion binding
alpha-actinin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200007653.mm.1 NM_001024618.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007653.mm.1 NM_001024618.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200007653.mm.1 NM_001083919.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007653.mm.1 NM_001083919.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007653.mm.1 TC0200000966.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200007654.mm.1 NM_001024618.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007654.mm.1 NM_001024618.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200007654.mm.1 NM_001083919.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007654.mm.1 NM_001083919.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007654.mm.1 TC0200000966.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0