This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01335498_g1
See other PCED1A GE Assays ›
Gene Symbol:
Pced1a
Gene Name:
PC-esterase domain containing 1A
Gene Aliases:
A930025D01Rik, AI480484, Fam113a
Chromosome Location:
Chr.2: 130417684 - 130424641 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01335498_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pced1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001114541.1 NP_001108013.1 2 - 3 287 68
NM_178762.4 NP_848877.2 1 - 2 874 68
GenBank mRNA AK032354.1 - 1 - 2 866 68
AK032762.1 - 1 - 2 849 68
AK045248.1 - 1 - 2 854 68
AK081584.1 - 1 - 2 835 68
AK161998.1 - 1 - 2 865 68
AK183067.1 - 1 - 2 88 68
AK183310.1 - 1 - 2 88 68
BC064802.1 - 2 - 3 333 30009464 68
BY326318.1 - 2 - 3 288 68

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK032354.1, AK032762.1, AK045248.1, AK081584.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pced1a

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200039567.mm.1 NM_001114541.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200039567.mm.1 NM_001114541.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200039567.mm.1 NM_178762.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200039567.mm.1 NM_178762.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200039567.mm.1 TC0200004626.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200039570.mm.1 NM_001114541.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200039570.mm.1 NM_001114541.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200039570.mm.1 NM_178762.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200039570.mm.1 NM_178762.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200039570.mm.1 TC0200004626.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200039571.mm.1 NM_001114541.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200039571.mm.1 NM_001114541.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200039571.mm.1 NM_178762.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200039571.mm.1 NM_178762.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200039571.mm.1 TC0200004626.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200039572.mm.1 NM_178762.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200039572.mm.1 NM_178762.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200039572.mm.1 TC0200004626.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200039580.mm.1 TC0200004626.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200039579.mm.1 NM_001114541.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200039579.mm.1 NM_001114541.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200039579.mm.1 TC0200004626.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 438784 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline