This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01335499_g1
See other PCED1A GE Assays ›
Gene Symbol:
Pced1a
Gene Name:
PC-esterase domain containing 1A
Gene Aliases:
A930025D01Rik, AI480484, Fam113a
Chromosome Location:
Chr.2: 130417684 - 130424641 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01335499_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pced1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001114541.1 NP_001108013.1 3 - 4 366 60
NM_178762.4 NP_848877.2 2 - 3 953 60
GenBank mRNA AK032354.1 - 2 - 3 945 60
AK032762.1 - 2 - 3 928 60
AK045248.1 - 2 - 3 933 60
AK085613.1 - 2 - 3 923 60
AK161998.1 - 2 - 3 944 60
BC064802.1 - 3 - 4 412 30009464 60

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK032354.1, AK032762.1, AK045248.1, AK085613.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pced1a

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200039565.mm.1 NM_001114541.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200039565.mm.1 NM_001114541.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200039565.mm.1 NM_178762.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200039565.mm.1 NM_178762.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200039565.mm.1 TC0200004626.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline