This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01335930_m1
See other CYBRD1 GE Assays ›
Gene Symbol:
Cybrd1
Gene Name:
cytochrome b reductase 1
Gene Aliases:
2210407P13Rik, Dcytb
Chromosome Location:
Chr.2: 71118054 - 71142926 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01335930_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cybrd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028593.2 NP_082869.2 2 - 3 461 73
GenBank mRNA AF354666.1 - 2 - 3 403 73
AK008849.1 - 2 - 3 407 73
AK029112.1 - 2 - 3 464 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001032.mm.1

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

transport
response to iron ion
oxidation-reduction process
ferric-chelate reductase activity
N-ethylmaleimide reductase activity
oxidoreductase activity
reduced coenzyme F420 dehydrogenase activity
sulfur oxygenase reductase activity
malolactic enzyme activity
NADPH:sulfur oxidoreductase activity
metal ion binding
epoxyqueuosine reductase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200008294.mm.1 NM_028593.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200008294.mm.1 NM_028593.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200008294.mm.1 TC0200001032.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 817954 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline