This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01336181_m1
See other SNAP25 GE Assays ›
Gene Symbol:
Snap25
Gene Name:
synaptosomal-associated protein 25
Gene Aliases:
Bdr, GENA70, SNAP-25, SUP, sp
Chromosome Location:
Chr.2: 136713438 - 136782428 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01336181_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Snap25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291056.1 NP_001277985.1 3 - 4 322 78
NM_011428.3 NP_035558.1 3 - 4 322 78
GenBank mRNA AF483516.1 - 2 - 3 109 78
AF483517.1 - 2 - 3 109 78
AK078038.1 - 3 - 4 78
AU080862.1 - 3 - 4 289 78
BC018249.1 - 3 - 4 210 4504644 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002077.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Snap25

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

exocytosis
neurotransmitter secretion
long-term memory
locomotory behavior
associative learning
regulation of neuron projection development
synaptic vesicle priming
endosomal transport
sleep
synaptic vesicle fusion to presynaptic active zone membrane
regulation of ion transmembrane transport
positive regulation of hormone secretion
calcium ion-regulated exocytosis of neurotransmitter
long-term synaptic potentiation
regulation of establishment of protein localization
potassium ion transmembrane transport
SNARE binding
voltage-gated potassium channel activity
SNAP receptor activity
protein binding
myosin binding
syntaxin-1 binding
protein domain specific binding
ion channel binding
protein N-terminus binding
calcium-dependent protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0