This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01336286_m1
See other ECHDC3 GE Assays ›
Gene Symbol:
Echdc3
Gene Name:
enoyl Coenzyme A hydratase domain containing 3
Gene Aliases:
2310005D12Rik, AI662097
Chromosome Location:
Chr.2: 6188465 - 6212994 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01336286_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Echdc3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024208.4 NP_077170.2 1 - 2 201 125
GenBank mRNA AK004218.1 - 1 - 2 125
AK009166.1 - 1 - 2 202 125
AK014736.1 - 1 - 2 229 125
BC002214.1 - 1 - 2 182 3490443 125
BC054365.1 - 1 - 2 170 5343451 125

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Echdc3

Gene Ontology Categories:

Function(s) Process(es)

biological_process
metabolic process
molecular_function
catalytic activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0