This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01336404_g1
See other NDOR1 GE Assays ›
Gene Symbol:
Ndor1
Gene Name:
NADPH dependent diflavin oxidoreductase 1
Gene Aliases:
4930447P04Rik, NR1, Ndor
Chromosome Location:
Chr.2: 25244813 - 25255414 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01336404_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndor1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001082476.2 NP_001075945.1 6 - 7 788 90
NM_001252541.1 NP_001239470.1 4 - 5 587 90
NM_001252542.1 NP_001239471.1 6 - 7 855 90
GenBank mRNA AK019617.1 - 6 - 7 785 90
AK143124.1 - 6 - 7 829 90
AK163207.1 - 4 - 5 557 90
AK169885.1 - 6 - 7 847 90
BC049789.1 - 6 - 7 838 6331033 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003140.mm.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

cell death
cellular response to menadione
oxidation-reduction process
NADPH-hemoprotein reductase activity
iron ion binding
FMN binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
epoxide carboxylase activity
flavin adenine dinucleotide binding
NADP binding
reduced coenzyme F420:NADP+ oxidoreductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200025836.mm.1 NM_001082476.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025836.mm.1 NM_001082476.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200025836.mm.1 NM_001252541.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025836.mm.1 NM_001252541.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025836.mm.1 NM_001252542.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025836.mm.1 NM_001252542.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025836.mm.1 TC0200003140.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200025839.mm.1 NM_001082476.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025839.mm.1 NM_001082476.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200025839.mm.1 NM_001252541.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025839.mm.1 NM_001252541.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025839.mm.1 NM_001252542.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025839.mm.1 NM_001252542.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025839.mm.1 TC0200003140.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200025837.mm.1 NM_001082476.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025837.mm.1 NM_001082476.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200025837.mm.1 NM_001252541.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025837.mm.1 NM_001252541.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025837.mm.1 NM_001252542.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025837.mm.1 NM_001252542.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025837.mm.1 TC0200003140.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200025838.mm.1 NM_001082476.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025838.mm.1 NM_001082476.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200025838.mm.1 NM_001252541.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025838.mm.1 NM_001252541.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025838.mm.1 NM_001252542.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025838.mm.1 NM_001252542.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200025838.mm.1 TC0200003140.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-9k2pj:80/100.66.79.197:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0