This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01336637_m1
See other SERAC1 GE Assays ›
Gene Symbol:
Serac1
Gene Name:
serine active site containing 1
Gene Aliases:
4930511N22Rik, D17Ertd141e
Chromosome Location:
Chr.17: 6040571 - 6079762 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01336637_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Serac1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001111017.1 NP_001104487.1 1 - 2 121 64
NM_177311.4 NP_796285.2 1 - 2 121 64
GenBank mRNA AK154605.1 - 1 - 2 72 64
AK155571.1 - 1 - 2 124 64
AK162246.1 - 1 - 2 56 64
AK170498.1 - 1 - 2 72 64
BC119594.1 - 1 - 2 110 40059066 64
BC119595.1 - 1 - 2 110 40059071 64
BY197974.1 - 1 - 2 121 64
BY715309.1 - 1 - 2 107 64

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK154605.1, AK155571.1, AK162246.1, AK170498.1, BC119594.1, BC119595.1, NM_001111017.1, NM_177311.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Serac1

Gene Ontology Categories:

Function(s) Process(es)

GPI anchor metabolic process
lipid metabolic process
intracellular protein transport
phospholipid biosynthetic process
extracellular matrix organization
hydrolase activity, acting on ester bonds

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700012407.mm.1 NM_001111017.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700012407.mm.1 NM_001111017.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700012407.mm.1 NM_177311.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700012407.mm.1 NM_177311.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700012407.mm.1 TC1700001378.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700012406.mm.1 NM_001111017.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700012406.mm.1 NM_001111017.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700012406.mm.1 NM_177311.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700012406.mm.1 NM_177311.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700012406.mm.1 TC1700001378.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 518914 - -
ABI Mouse Genome Survey Microarray 772618 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0