This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01336641_m1
See other SERAC1 GE Assays ›
Gene Symbol:
Serac1
Gene Name:
serine active site containing 1
Gene Aliases:
4930511N22Rik, D17Ertd141e
Chromosome Location:
Chr.17: 6040571 - 6079762 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01336641_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Serac1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001111017.1 NP_001104487.1 9 - 10 967 80
NM_177311.4 NP_796285.2 8 - 9 877 80
GenBank mRNA AK019677.1 - 8 - 9 866 80
AK154605.1 - 9 - 10 918 80
AK155571.1 - 9 - 10 970 80
AK162246.1 - 9 - 10 902 80
AK170498.1 - 9 - 10 918 80
BC119594.1 - 9 - 10 956 40059066 80
BC119595.1 - 9 - 10 956 40059071 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001378.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Serac1

Gene Ontology Categories:

Function(s) Process(es)

GPI anchor metabolic process
lipid metabolic process
intracellular protein transport
phospholipid biosynthetic process
extracellular matrix organization
hydrolase activity, acting on ester bonds

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700012398.mm.1 NM_001111017.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700012398.mm.1 NM_001111017.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700012398.mm.1 NM_177311.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700012398.mm.1 NM_177311.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700012398.mm.1 TC1700001378.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2d9fq:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0