This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01336797_m1
See other NDUFA8 GE Assays ›
Gene Symbol:
Ndufa8
Gene Name:
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 8
Gene Aliases:
0610033L03Rik, AW261656
Chromosome Location:
Chr.2: 36036334 - 36049292 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01336797_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufa8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026703.2 NP_080979.1 3 - 4 433 105
GenBank mRNA AK002744.1 - 3 - 4 433 105
AK088463.1 - 3 - 4 479 105
BC012416.1 - 3 - 4 431 3584066 105
BC024673.1 - 3 - 4 430 4195454 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003376.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndufa8

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
oxidation-reduction process
protein complex binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21137234 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21137236 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21137436 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21137437 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21137237 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21137235 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21137435 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475501 NM_026703.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475501 AK002744.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475501 AK088463.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475501 BC012416.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475501 BC024673.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475501 NM_026703.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475501 AK002744.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475501 AK088463.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475501 BC012416.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P475501 BC024673.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 878879 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490373 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579683 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 758195 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 289822 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626976 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 87511 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 32827 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117906 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160145 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475501 NM_026703.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475501 AK002744.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475501 AK088463.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475501 BC012416.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475501 BC024673.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475501 NM_026703.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475501 AK002744.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475501 AK088463.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475501 BC012416.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P475501 BC024673.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P475501 NM_026703.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P475501 AK002744.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P475501 AK088463.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P475501 BC012416.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P475501 BC024673.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P475501 NM_026703.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P475501 AK002744.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P475501 AK088463.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P475501 BC012416.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P475501 BC024673.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137413 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137418 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137409 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137424 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137425 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137428 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137432 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137427 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137420 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137434 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137402 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137407 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137400 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137411 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137422 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137415 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137419 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137417 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137435 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21137405 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04485917 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117906 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160145 - -
Whole Mouse Genome G4122A A_51_P475501 NM_026703.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P475501 AK002744.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P475501 AK088463.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P475501 BC012416.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P475501 BC024673.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P475501 NM_026703.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P475501 AK002744.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P475501 AK088463.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P475501 BC012416.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P475501 BC024673.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132805 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160145 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117906 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02124240 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02124249 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02124247 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02124248 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21137437 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21137235 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21137435 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21137234 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21137236 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21137237 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21137436 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137363 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137400 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137358 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137434 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137376 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137338 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137313 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137345 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137416 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137360 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137357 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137372 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137409 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137361 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137368 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137396 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137301 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137354 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137319 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137423 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137392 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137383 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137353 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137389 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137377 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137348 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137341 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137333 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137303 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137411 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137373 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137344 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137322 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137432 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137311 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137405 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137428 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137407 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137355 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137359 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137346 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137314 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137330 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137375 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137422 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137398 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137339 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137402 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137299 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137419 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137335 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137351 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137309 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137326 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137380 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137316 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137366 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137307 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137435 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137427 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137413 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137370 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137425 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137430 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137343 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137384 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21137312 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01510846 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01573181 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137303 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137402 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137326 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137405 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137357 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137344 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137343 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01500075 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01904693 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01427752 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137425 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137432 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01500076 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02212017 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137434 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137396 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137411 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137422 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01573186 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137429 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01879721 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137335 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137301 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137304 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137299 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137300 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137305 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137413 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01573183 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137339 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02024213 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137346 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137417 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137341 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02182194 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02024209 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01510847 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137348 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137318 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137435 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01209534 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137427 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01510850 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01427760 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137351 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137355 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02024212 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02145394 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137407 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137328 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137389 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137331 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137392 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137394 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01997920 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137358 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137400 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137333 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137430 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21137338 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10482092 - -
Affymetrix Mouse Gene 1.0 ST Array 10482094 - -
Affymetrix Mouse Gene 1.0 ST Array 10471705 - -
Affymetrix Mouse Gene 1.0 ST Array 10482090 - -
Affymetrix Mouse Gene 1.0 ST Array 10482091 - -
Affymetrix HT MG-430 PM Array Plate 1423692_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10482089 - -
Affymetrix Mouse Gene 1.1 ST Array 10471703 - -
Mouse Genome 430 2.0 Array 1423692_PM_at - -
Mouse Genome 430 2.0 Array 1423692_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 68375_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 68375_PM_at - -
Affymetrix Mouse Clariom S Array PSR0200029038.mm.1 NM_026703.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200029038.mm.1 NM_026703.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200029038.mm.1 TC0200003376.mm.1 MTA TC
Mouse Expression Set 430 1423692_at - -
Mouse Expression Set 430 1423692_PM_at - -
Murine Genome U74v2 Set 93531_at - -
Murine Genome U74v2 Set 93531_PM_at - -
Mouse Genome 430A 2.0 Array 1423692_PM_at - -
Mouse Genome 430A 2.0 Array 1423692_at - -
Affymetrix HT Mouse Genome 430A Array 1423692_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1423692_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 924067 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 5050242 - -
Illumina mouse-6 v1.0 expression beadchip 5050242 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005050242 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2682019 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2682019 - -
mouse-6 v1.1 (Illumina) scl19390.3.3_3-S - -
Illumina Sentrix mouse-6.v1 bead array 5050242 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl19390.3.3_3-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5050242 - -
MouseRef-8 v2 Expression BeadChip ILMN_2880067 - -
MouseRef-8 v2 Expression BeadChip ILMN_2682019 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2682019 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2880067 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl19390.3.3_3-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5bwlp:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0