This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01336944_m1
See other GFI1B GE Assays ›
Gene Symbol:
Gfi1b
Gene Name:
growth factor independent 1B
Gene Aliases:
Gfi-1B
Chromosome Location:
Chr.2: 28609450 - 28621982 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01336944_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gfi1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160406.1 NP_001153878.1 1 - 2 130 78
NM_008114.3 NP_032140.1 1 - 2 130 78
GenBank mRNA AK157262.1 - 1 - 2 139 78
BC052654.1 - 1 - 2 105 30039774 78
BC132365.1 - 1 - 2 66 40130642 78
BC132367.1 - 1 - 2 66 40130644 78
BC145238.1 - 1 - 2 66 9053790 78
W65842.1 - 1 - 2 130 77

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK157262.1, BC052654.1, BC132365.1, BC132367.1, BC145238.1, NM_001160406.1, NM_008114.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gfi1b

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
hemopoiesis
myeloid cell differentiation
positive regulation of granulocyte differentiation
regulation of erythrocyte differentiation
regulation of neurogenesis
regulation of histone H3-K4 methylation
negative regulation of histone H3-K4 methylation
positive regulation of histone H3-K9 methylation
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
RNA polymerase II transcription factor binding
nucleic acid binding
DNA binding
protein binding
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200027291.mm.1 NM_001160406.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027291.mm.1 NM_001160406.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027291.mm.1 NM_008114.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027291.mm.1 NM_008114.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200027291.mm.1 TC0200003234.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200027293.mm.1 NM_001160406.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027293.mm.1 NM_001160406.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027293.mm.1 NM_008114.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027293.mm.1 NM_008114.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200027293.mm.1 TC0200003234.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200027295.mm.1 NM_001160406.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027295.mm.1 NM_001160406.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027295.mm.1 NM_008114.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027295.mm.1 NM_008114.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200027295.mm.1 TC0200003234.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200027294.mm.1 NM_001160406.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027294.mm.1 NM_001160406.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027294.mm.1 NM_008114.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027294.mm.1 NM_008114.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200027294.mm.1 TC0200003234.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 766109 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0