This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01337111_g1
See other PTPN21 GE Assays ›
Gene Symbol:
Ptpn21
Gene Name:
protein tyrosine phosphatase, non-receptor type 21
Gene Aliases:
PTPD1, PTPRL10
Chromosome Location:
Chr.12: 98676741 - 98737618 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01337111_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptpn21
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001146199.1 NP_001139671.1 17 - 18 3467 69
NM_011877.2 NP_036007.2 17 - 18 3602 69
GenBank mRNA BC049893.1 - 15 - 16 3349 69
BC055942.1 - 5 - 6 1991 69
BC098500.1 - 17 - 18 3467 30081068 69
D37801.1 - 17 - 18 3551 69
D83072.1 - 4 - 5 863 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200002194.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptpn21

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
dephosphorylation
peptidyl-tyrosine dephosphorylation
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
hydrolase activity
phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200015139.mm.1 NM_001146199.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200015139.mm.1 NM_001146199.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200015139.mm.1 NM_011877.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200015139.mm.1 NM_011877.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200015139.mm.1 TC1200002194.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0