This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01337457_g1
See other IDI1 GE Assays ›
Gene Symbol:
Idi1
Gene Name:
isopentenyl-diphosphate delta isomerase
Gene Aliases:
4832416K17Rik
Chromosome Location:
Chr.13: 8885606 - 8892396 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01337457_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Idi1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145360.2 NP_663335.2 4 - 5 787 94
GenBank mRNA AK029302.1 - 4 - 5 420 94
AK084208.1 - 4 - 5 418 94
AK160832.1 - 4 - 5 421 94
BC004801.1 - 3 - 4 408 3589498 94
BC110313.1 - 4 - 5 425 5343431 94
CT010310.1 - 3 - 4 365 94

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Idi1

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

isoprenoid biosynthetic process
magnesium ion binding
isopentenyl-diphosphate delta-isomerase activity
manganese ion binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028961 BC004801.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028961 BC110313.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028961 BE953932.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028961 AK132073.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028961 AK160832.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121772 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028961 BC004801.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028961 BC110313.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028961 BE953932.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028961 AK132073.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028961 AK160832.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28813475 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28813482 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28813476 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28813479 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28813483 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28813466 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28813468 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28813473 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28813464 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28813469 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28813465 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28813472 - -
Whole Mouse Genome G4122A A_51_P329711 BC004801.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P329711 BC110313.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P329711 BE953932.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P329711 AK160832.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P329711 NM_145360.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P441634 BC004801.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P441634 BC110313.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P441634 CT010310.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P441634 AK160832.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P441634 NM_145360.2 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113506 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10499574 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10499566 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10499577 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10499568 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10499573 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813487 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813426 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813486 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813475 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813476 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813511 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813468 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813423 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813505 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813469 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813471 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813466 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813483 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813472 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813484 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813479 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813482 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813506 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28813514 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28813483 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28813479 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28813473 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10499573 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28813476 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11061466 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28813469 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28813468 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28813482 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28813475 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28813466 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11061463 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28813471 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28813510 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28813505 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28813472 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28813488 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28813465 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28813486 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28813484 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10723927 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10602963 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10482763 - -
Affymetrix Mouse Gene 1.0 ST Array 10403415 - -
Affymetrix Mouse Gene 1.0 ST Array 10482765 - -
Affymetrix Mouse Gene 1.0 ST Array 10403418 - -
Affymetrix Mouse Gene 1.0 ST Array 10403416 - -
Affymetrix Mouse Gene 1.0 ST Array 10403414 - -
Affymetrix Mouse Gene 1.0 ST Array 10403419 - -
Affymetrix Mouse Gene 1.0 ST Array 10482764 - -
Affymetrix Mouse Gene 1.0 ST Array 10403417 - -
Affymetrix HT MG-430 PM Array Plate 1451122_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1423804_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10403413 - -
Affymetrix Mouse Gene 1.1 ST Array 10482762 - -
Mouse Genome 430 2.0 Array 1451122_PM_at - -
Mouse Genome 430 2.0 Array 1423804_PM_a_at - -
Mouse Genome 430 2.0 Array 1423804_a_at - -
Mouse Genome 430 2.0 Array 1451122_at - -
Affymetrix Mouse Clariom S Array PSR1300000315.mm.1 NM_145360.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300000315.mm.1 NM_145360.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300000315.mm.1 TC1300000045.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300000316.mm.1 NM_145360.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300000316.mm.1 NM_145360.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300000316.mm.1 TC1300000045.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300000317.mm.1 NM_145360.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300000317.mm.1 NM_145360.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300000317.mm.1 TC1300000045.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300000318.mm.1 NM_145360.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300000318.mm.1 NM_145360.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300000318.mm.1 TC1300000045.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300000313.mm.1 NM_145360.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300000313.mm.1 NM_145360.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300000313.mm.1 TC1300000045.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300000312.mm.1 NM_145360.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300000312.mm.1 NM_145360.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300000312.mm.1 TC1300000045.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300000311.mm.1 NM_145360.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300000311.mm.1 NM_145360.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300000311.mm.1 TC1300000045.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300000314.mm.1 NM_145360.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300000314.mm.1 NM_145360.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300000314.mm.1 TC1300000045.mm.1 MTA TC
Mouse Expression Set 430 1423804_a_at - -
Mouse Expression Set 430 1423804_PM_a_at - -
Mouse Expression Set 430 1451122_at - -
Mouse Expression Set 430 1451122_PM_at - -
Murine Genome U74v2 Set 96269_PM_at - -
Murine Genome U74v2 Set 96269_at - -
Mouse Genome 430A 2.0 Array 1451122_PM_at - -
Mouse Genome 430A 2.0 Array 1423804_a_at - -
Mouse Genome 430A 2.0 Array 1451122_at - -
Mouse Genome 430A 2.0 Array 1423804_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1423804_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1451122_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1423804_a_at - -
Affymetrix HT Mouse Genome 430A Array 1451122_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 1450736 - -
Illumina mouse-6 v1.0 expression beadchip 1450736 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001450736 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2590923 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2590923 - -
mouse-6 v1.1 (Illumina) scl0319554.7_300-S - -
Illumina Sentrix mouse-6.v1 bead array 1450736 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0319554.7_300-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1450736 - -
MouseRef-8 v2 Expression BeadChip ILMN_2590923 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2590923 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline