This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01337915_g1
See other SPINT1 GE Assays ›
Gene Symbol:
Spint1
Gene Name:
serine protease inhibitor, Kunitz type 1
Gene Aliases:
HAI-1
Chromosome Location:
Chr.2: 119237360 - 119249535 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01337915_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Spint1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016907.3 NP_058603.2 4 - 5 922 71
GenBank mRNA AK017342.1 - 3 - 4 804 71
BC005769.1 - 3 - 4 809 2650110 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001765.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Spint1

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

neural tube closure
embryonic placenta development
negative regulation of peptidase activity
extracellular matrix organization
branching involved in labyrinthine layer morphogenesis
placenta blood vessel development
serine-type endopeptidase inhibitor activity
peptidase inhibitor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200013216.mm.1 NM_016907.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200013216.mm.1 NM_016907.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200013216.mm.1 TC0200001765.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline