This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01337994_m1
See other BMF GE Assays ›
Gene Symbol:
Bmf
Gene Name:
BCL2 modifying factor
Gene Aliases:
AW260063, AW411872
Chromosome Location:
Chr.2: 118528757 - 118549678 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01337994_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bmf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138313.3 NP_612186.2 2 - 3 267 65
GenBank mRNA AK040622.1 - 2 - 3 224 65
AK084658.1 - 2 - 3 277 65
AK169903.1 - 2 - 3 224 65
BC079650.1 - 2 - 3 281 6847314 65

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK040622.1, BC079650.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bmf

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
positive regulation of protein homooligomerization
anoikis
positive regulation of release of cytochrome c from mitochondria
negative regulation of apoptotic signaling pathway
positive regulation of apoptotic signaling pathway
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200036819.mm.1 NM_138313.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200036819.mm.1 TC0200004415.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200036820.mm.1 NM_138313.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200036820.mm.1 TC0200004415.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lsr54:80/100.66.79.61:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0