This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01338234_m1
See other NOSTRIN GE Assays ›
Gene Symbol:
Nostrin
Gene Name:
nitric oxide synthase trafficker
Gene Aliases:
Daip2, mDaIP2
Chromosome Location:
Chr.2: 69135800 - 69189330 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01338234_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nostrin
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181547.3 NP_853525.3 5 - 6 434 100
GenBank mRNA AJ564264.1 - 5 - 6 344 100
AK052926.1 - 5 - 6 426 100
AY280963.1 - 5 - 6 387 100
BC069942.1 - 5 - 6 411 30443942 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001002.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nostrin

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
negative regulation of transcription, DNA-templated
regulation of nitric-oxide synthase activity
DNA binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200007834.mm.1 NM_181547.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007834.mm.1 NM_181547.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200007834.mm.1 TC0200001002.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200007833.mm.1 NM_181547.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200007833.mm.1 NM_181547.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200007833.mm.1 TC0200001002.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0