This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01338504_m1
See other CDYL GE Assays ›
Gene Symbol:
Cdyl
Gene Name:
chromodomain protein, Y chromosome-like
Gene Aliases:
AI325931
Chromosome Location:
Chr.13: 35659811 - 35874064 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01338504_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdyl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009881.3 NP_034011.1 2 - 3 270 70
GenBank mRNA AF081260.1 - 2 - 3 252 70
AF081261.1 - 2 - 3 252 70
BC055103.1 - 2 - 3 219 6741468 70
BY090531.1 - 2 - 3 271 70

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF081260.1, AF081261.1, BC055103.1, NM_009881.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdyl

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
metabolic process
negative regulation of nucleic acid-templated transcription
transcription corepressor activity
catalytic activity
histone acetyltransferase activity
transferase activity
transferase activity, transferring acyl groups
methylated histone binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300002445.mm.1 NM_009881.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300002445.mm.1 NM_009881.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300002445.mm.1 TC1300000405.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300002446.mm.1 NM_009881.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300002446.mm.1 NM_009881.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300002446.mm.1 TC1300000405.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300002443.mm.1 NM_009881.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300002443.mm.1 NM_009881.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300002443.mm.1 TC1300000405.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300002444.mm.1 NM_009881.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300002444.mm.1 NM_009881.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300002444.mm.1 TC1300000405.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 455218 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0