This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01339771_m1
See other STARD7 GE Assays ›
Gene Symbol:
Stard7
Gene Name:
START domain containing 7
Gene Aliases:
AI852671, AL022671, AW544915
Chromosome Location:
Chr.2: 127270203 - 127298935 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01339771_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stard7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_139308.2 NP_647469.2 6 - 7 1203 80
GenBank mRNA AF244543.1 - 5 - 6 561 80
AK083125.1 - 5 - 6 958 80
AK143699.1 - 6 - 7 1204 80
AK182956.1 - 1 - 2 80
BC017524.1 - 6 - 7 1193 4527790 80
BC080747.1 - 6 - 7 1174 4237000 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001911.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stard7

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein targeting to mitochondrion
molecular_function
lipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200014979.mm.1 NM_139308.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200014979.mm.1 NM_139308.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200014979.mm.1 TC0200001911.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0