This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01339787_m1
See other PPP6C GE Assays ›
Gene Symbol:
Ppp6c
Gene Name:
protein phosphatase 6, catalytic subunit
Gene Aliases:
2310003C10Rik
Chromosome Location:
Chr.2: 39194369 - 39226338 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01339787_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp6c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024209.2 NP_077171.1 3 - 4 330 132
GenBank mRNA AK002764.1 - 3 - 4 292 132
AK009104.1 - 3 - 4 310 132
BC002223.1 - 3 - 4 355 3491111 132

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003440.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp6c

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
cell cycle
phosphoprotein phosphatase activity
protein serine/threonine phosphatase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200029516.mm.1 NM_024209.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200029516.mm.1 NM_024209.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200029516.mm.1 TC0200003440.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200029515.mm.1 NM_024209.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200029515.mm.1 NM_024209.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200029515.mm.1 TC0200003440.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0