This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01339801_m1
See other SLC22A23 GE Assays ›
Gene Symbol:
Slc22a23
Gene Name:
solute carrier family 22, member 23
Gene Aliases:
3110004L20Rik
Chromosome Location:
Chr.13: 34179158 - 34345487 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01339801_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc22a23
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033167.3 NP_001028339.1 5 - 6 1577 65
GenBank mRNA AK147397.1 - 5 - 6 1578 65
BC039219.1 - 2 - 3 146 65
BC041671.1 - 2 - 3 146 65
BC053705.1 - 3 - 4 365 65
BC094372.1 - 46 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300001784.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
transmembrane transport
molecular_function
transporter activity
transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300011145.mm.1 NM_001033167.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300011145.mm.1 NM_001033167.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300011145.mm.1 TC1300001784.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0