This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01340182_g1
See other RBM17 GE Assays ›
Gene Symbol:
Rbm17
Gene Name:
RNA binding motif protein 17
Gene Aliases:
2700027J02Rik
Chromosome Location:
Chr.2: 11585439 - 11603199 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01340182_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbm17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_152824.1 NP_690037.1 2 - 3 252 60
GenBank mRNA AF083384.1 - 1 - 2 122 60
AK012296.1 - 2 - 3 196 60
BC034896.1 - 2 - 3 252 1446602 60

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbm17

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

RNA splicing, via transesterification reactions
alternative mRNA splicing, via spliceosome
mRNA processing
RNA splicing
nucleotide binding
nucleic acid binding
RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200024409.mm.1 NM_152824.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200024409.mm.1 NM_152824.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200024409.mm.1 TC0200002963.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200024410.mm.1 NM_152824.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200024410.mm.1 NM_152824.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200024410.mm.1 TC0200002963.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200024411.mm.1 NM_152824.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200024411.mm.1 NM_152824.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200024411.mm.1 TC0200002963.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200024408.mm.1 NM_152824.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200024408.mm.1 NM_152824.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200024408.mm.1 TC0200002963.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rz6mw:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0