This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01340867_m1
See other GLA GE Assays ›
Gene Symbol:
Gla
Gene Name:
galactosidase, alpha
Gene Aliases:
Ags
Chromosome Location:
Chr.X: 134588169 - 134601005 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01340867_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gla
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013463.2 NP_038491.2 4 - 5 709 151
GenBank mRNA AK040081.1 - 4 - 5 641 151
AK054547.1 - 4 - 5 661 151
AK164231.1 - 4 - 5 642 151
BC009021.1 - 4 - 5 662 3708950 151
U34071.1 - 4 - 5 659 151
U50716.1 - 4 - 5 647 151

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00002995.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gla

Panther Classification:

Molecular Function -

galactosidase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
metabolic process
oligosaccharide metabolic process
glycoside catabolic process
bone mineralization
negative regulation of nitric oxide biosynthetic process
glycosylceramide catabolic process
negative regulation of nitric-oxide synthase activity
catalytic activity
hydrolase activity, hydrolyzing O-glycosyl compounds
alpha-galactosidase activity
receptor binding
hydrolase activity
hydrolase activity, acting on glycosyl bonds
galactoside binding
protein homodimerization activity
raffinose alpha-galactosidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00018692.mm.1 NM_013463.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00018692.mm.1 NM_013463.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00018692.mm.1 TC0X00002995.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0