This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01340869_m1
See other GLA GE Assays ›
Gene Symbol:
Gla
Gene Name:
galactosidase, alpha
Gene Aliases:
Ags
Chromosome Location:
Chr.X: 134588169 - 134601005 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01340869_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gla
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013463.2 NP_038491.2 6 - 7 1075 58
GenBank mRNA AK040081.1 - 6 - 7 1007 58
AK054547.1 - 6 - 7 1027 58
AK145011.1 - 6 - 7 1017 58
AK145148.1 - 1075 58
AK164231.1 - 6 - 7 1008 58
AK166204.1 - 6 - 7 1008 58
AK166245.1 - 6 - 7 1024 58
BC009021.1 - 6 - 7 1028 3708950 58
U34071.1 - 6 - 7 1025 58
U50716.1 - 6 - 7 1013 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00002995.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gla

Panther Classification:

Molecular Function -

galactosidase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
metabolic process
oligosaccharide metabolic process
glycoside catabolic process
bone mineralization
negative regulation of nitric oxide biosynthetic process
glycosylceramide catabolic process
negative regulation of nitric-oxide synthase activity
catalytic activity
hydrolase activity, hydrolyzing O-glycosyl compounds
alpha-galactosidase activity
receptor binding
hydrolase activity
hydrolase activity, acting on glycosyl bonds
galactoside binding
protein homodimerization activity
raffinose alpha-galactosidase activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672780 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672781 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2146136 U34071.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2146136 AK145148.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2146136 AK164231.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2146136 AK040081.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2146136 NM_013463.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48830 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 518202 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 371693 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540293 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 14224 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633806 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 932472 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973547 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921033 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203784 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 108501 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 4375 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155008 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107866 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2146136 U34071.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2146136 AK145148.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2146136 AK164231.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2146136 AK040081.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2146136 NM_013463.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P153265 U34071.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P234073 U34071.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P234073 U50716.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P234073 AK166204.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P234073 AK166245.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P234073 BC009021.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672765 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672766 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672775 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672781 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672756 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672760 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672748 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672776 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672759 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672742 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672772 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672751 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672754 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672749 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672778 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672770 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672743 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672767 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672746 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672779 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672744 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672763 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672747 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672755 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672771 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672769 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672752 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672741 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672774 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672758 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09426132 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09426140 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107866 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03757948 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155008 - -
Whole Mouse Genome G4122A A_52_P234073 U34071.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P234073 U50716.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P234073 AK166204.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P234073 AK166245.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P234073 BC009021.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P153265 U34071.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109784 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108697 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107866 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155008 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14846952 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14846928 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14846956 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14846932 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14846941 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14846944 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672781 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672780 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672755 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672733 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672686 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672723 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672738 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672645 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672677 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672751 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672742 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672713 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672638 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672691 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672695 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672730 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672641 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672746 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672739 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672778 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672721 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672709 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672752 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672673 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672681 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672734 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672654 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672650 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672684 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672748 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672774 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672648 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672653 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672743 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672761 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672717 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672772 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672754 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672760 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672725 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672749 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672758 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672727 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672698 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672763 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672769 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672706 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672718 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672661 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672724 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672647 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672652 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672715 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672765 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672767 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672690 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672710 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672728 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672744 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672657 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672771 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672707 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672699 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672775 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672693 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672781 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672668 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672735 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672636 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672656 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672732 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672779 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672644 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672701 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672678 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672679 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672660 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672676 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672711 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672664 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672736 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672719 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672756 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672740 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672682 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672759 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672672 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672687 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672689 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672770 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672659 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672723 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672743 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672732 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14810589 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15112060 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672739 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672771 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672779 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14992234 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672755 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672765 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15060122 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672704 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672709 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672734 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672742 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672718 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672733 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672770 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672728 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672727 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15060120 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672753 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672767 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672763 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672705 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672736 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15129569 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672761 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672707 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672754 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672774 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672715 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672756 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15129562 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672759 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672724 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672725 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672747 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672758 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672746 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672772 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15180194 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672744 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672760 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672751 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672775 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672748 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672778 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672721 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672719 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672711 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672717 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672749 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32672730 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10606722 - -
Affymetrix Mouse Gene 1.0 ST Array 10606723 - -
Affymetrix Mouse Gene 1.0 ST Array 10606717 - -
Affymetrix Mouse Gene 1.0 ST Array 10606721 - -
Affymetrix Mouse Gene 1.0 ST Array 10606715 - -
Affymetrix Mouse Gene 1.0 ST Array 10606716 - -
Affymetrix Mouse Gene 1.0 ST Array 10606720 - -
Affymetrix Mouse Gene 1.0 ST Array 10606718 - -
Affymetrix Mouse Gene 1.0 ST Array 10606724 - -
Affymetrix Mouse Gene 1.0 ST Array 10606719 - -
Affymetrix HT MG-430 PM Array Plate 1459738_PM_x_at - -
Affymetrix HT MG-430 PM Array Plate 1449006_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1418248_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10606714 - -
Mouse Genome 430 2.0 Array 1449006_PM_at - -
Mouse Genome 430 2.0 Array 1418248_at - -
Mouse Genome 430 2.0 Array 1418248_PM_at - -
Mouse Genome 430 2.0 Array 1459738_x_at - -
Mouse Genome 430 2.0 Array 1459738_PM_x_at - -
Mouse Genome 430 2.0 Array 1449006_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 11605_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 11605_PM_at - -
Affymetrix Murine Genome 6500A Array Msa16100_at - -
Affymetrix Mouse Clariom S Array PSR0X00018689.mm.1 NM_013463.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00018689.mm.1 NM_013463.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00018689.mm.1 TC0X00002995.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00018690.mm.1 NM_013463.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00018690.mm.1 NM_013463.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00018690.mm.1 TC0X00002995.mm.1 MTA TC
Mouse Expression Set 430 1449006_at - -
Mouse Expression Set 430 1418248_PM_at - -
Mouse Expression Set 430 1418248_at - -
Mouse Expression Set 430 1449006_PM_at - -
Mouse Expression Set 430 1459738_x_at - -
Mouse Expression Set 430 1459738_PM_x_at - -
Murine Genome U74v2 Set 102341_PM_at - -
Murine Genome U74v2 Set 102341_at - -
Mouse Genome 430A 2.0 Array 1449006_at - -
Mouse Genome 430A 2.0 Array 1418248_at - -
Mouse Genome 430A 2.0 Array 1418248_PM_at - -
Mouse Genome 430A 2.0 Array 1449006_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1418248_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1449006_at - -
Affymetrix HT Mouse Genome 430A Array 1418248_at - -
Affymetrix HT Mouse Genome 430A Array 1449006_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1459738_x_at - -
Affymetrix HT Mouse Genome 430B Array 1459738_PM_x_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 102370731 - -
Illumina mouse-6 v1.0 expression beadchip 4610364 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004610364 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2626086 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1220292 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2626086 - -
mouse-6 v1.1 (Illumina) scl0011605.2_12-S - -
mouse-6 v1.1 (Illumina) scl53832.8_267-S - -
Illumina Sentrix mouse-6.v1 bead array 102370731 - -
Illumina Sentrix mouse-6.v1 bead array 4610364 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0011605.2_12-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4610364 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 102370731 - -
MouseRef-8 v2 Expression BeadChip ILMN_2626086 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2626086 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0