This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01341197_m1
See other PALLD GE Assays ›
Gene Symbol:
Palld
Gene Name:
palladin, cytoskeletal associated protein
Gene Aliases:
2410003B16Rik, 6030492A02
Chromosome Location:
Chr.8: 61511429 - 61902700 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01341197_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Palld
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001293772.1 NP_001280701.1 11 - 12 2889 68
NM_001293773.1 NP_001280702.1 10 - 11 2020 68
NM_001293774.1 NP_001280703.1 2 - 3 786 68
GenBank mRNA AK031696.1 - 10 - 11 2012 68
AK052489.1 - 11 - 12 2872 68
AK076423.1 - 9 - 10 1658 68
AK149663.1 - 2 - 3 762 68
AK173081.1 - 10 - 11 1960 68
BC027364.1 - 10 - 11 1969 68
BC076588.1 - 3 - 4 1048 68
BC127081.1 - 2 - 3 743 68
JX477684.1 - 10 - 11 2012 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

keratinocyte development
epithelial cell morphogenesis
actin cytoskeleton organization
neuron projection development
ruffle organization
positive regulation of podosome assembly
actin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0