This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01341198_m1
See other PALLD GE Assays ›
Gene Symbol:
Palld
Gene Name:
palladin, cytoskeletal associated protein
Gene Aliases:
2410003B16Rik, 6030492A02
Chromosome Location:
Chr.8: 61511429 - 61902700 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01341198_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Palld
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081390.1 NP_001074859.1 11 - 12 2348 88
NM_001293772.1 NP_001280701.1 12 - 13 3027 88
NM_001293773.1 NP_001280702.1 11 - 12 2158 88
NM_001293774.1 NP_001280703.1 3 - 4 924 88
GenBank mRNA AF205078.1 - 1 - 2 102 88
AK031696.1 - 11 - 12 2150 88
AK052489.1 - 12 - 13 3010 88
AK076423.1 - 10 - 11 1796 88
AK087573.1 - 3 - 4 300 88
AK149663.1 - 3 - 4 900 88
AK173081.1 - 11 - 12 2098 88
BC027364.1 - 11 - 12 2107 88
BC076588.1 - 4 - 5 1186 88
BC127081.1 - 3 - 4 881 88
JX477684.1 - 11 - 12 2150 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002300.mm.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

keratinocyte development
epithelial cell morphogenesis
actin cytoskeleton organization
neuron projection development
ruffle organization
positive regulation of podosome assembly
actin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800017344.mm.1 NM_001081390.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800017344.mm.1 NM_001081390.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800017344.mm.1 TC0800002300.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800017343.mm.1 NM_001081390.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800017343.mm.1 NM_001081390.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800017343.mm.1 TC0800002300.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0