This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01341203_m1
See other PALLD GE Assays ›
Gene Symbol:
Palld
Gene Name:
palladin, cytoskeletal associated protein
Gene Aliases:
2410003B16Rik, 6030492A02
Chromosome Location:
Chr.8: 61511429 - 61902700 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01341203_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Palld
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081390.1 NP_001074859.1 16 - 17 3042 83
NM_001293772.1 NP_001280701.1 18 - 19 3772 83
NM_001293773.1 NP_001280702.1 16 - 17 2852 83
NM_001293774.1 NP_001280703.1 8 - 9 1618 83
GenBank mRNA AF205078.1 - 6 - 7 796 83
AK010350.1 - 4 - 5 505 83
AK031696.1 - 16 - 17 2844 83
AK052489.1 - 18 - 19 3755 83
AK076423.1 - 15 - 16 2490 83
AK087573.1 - 8 - 9 994 83
AK149663.1 - 8 - 9 1595 83
AK173081.1 - 16 - 17 2792 83
BC027364.1 - 16 - 17 2801 83
BC076588.1 - 9 - 10 1880 83
BC127081.1 - 9 - 10 1626 83
JX477684.1 - 16 - 17 2844 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

keratinocyte development
epithelial cell morphogenesis
actin cytoskeleton organization
neuron projection development
ruffle organization
positive regulation of podosome assembly
actin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0