This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01341950_m1
See other HERC6 GE Assays ›
Gene Symbol:
Herc6
Gene Name:
hect domain and RLD 6
Gene Aliases:
1700121D12Rik, 2510038N07Rik, 4930427L17Rik, AI451296, CEB1, Herc5
Chromosome Location:
Chr.6: 57580992 - 57665136 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01341950_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Herc6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025992.2 NP_080268.1 17 - 18 2253 83
GenBank mRNA AK147960.1 - 17 - 18 2176 83
AK149648.1 - 17 - 18 2245 83
AK161293.1 - 5 - 6 859 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600003503.mm.1

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
immune system process
protein ubiquitination
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
innate immune response
ubiquitin-protein transferase activity
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
cyclin binding
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P64514 AK149648.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P64514 AK147960.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P64514 AK011047.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P64514 AK015214.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P64514 AK017888.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P64514 AK149648.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P64514 AK147960.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P64514 AK011047.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P64514 AK015214.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P64514 AK017888.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P679860 AK147960.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P679860 NM_025992.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P679860 AK161293.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P679860 AK147960.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P679860 NM_025992.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P679860 AK161293.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 922699 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 882468 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 769305 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 416645 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 185722 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 734053 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 761614 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 497795 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 672177 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 272985 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 811498 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900658 BC092392.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900658 NM_001099308.1 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900658 NM_001099327.1 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900658 NM_001099349.1 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900658 NM_001100415.1 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900658 NM_001100416.1 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900658 NM_001101804.1 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900658 NM_001113806.1 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900658 XM_001004273.2 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900658 XM_001472096.1 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900658 XM_001472180.1 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900658 XM_001480607.1 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900658 XM_001480913.1 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900658 XM_001481229.1 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900658 XM_001481230.1 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900658 XM_001481339.1 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900658 XM_902322.3 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 100102 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648977 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 934681 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 610769 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724519 AK007899.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 917357 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 135745 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 521485 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 697127 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 546569 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969805 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 608216 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 948100 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 363830 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284357 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 715352 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 866252 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 296366 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 274450 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 400986 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924012 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 134660 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 611471 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 412306 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 436535 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 620395 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 646702 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 446135 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 105309 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240442 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 145328 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632052 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 590251 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 73717 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 141664 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 81691 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 69170 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 152270 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 171659 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13560 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 39281 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109368 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105987 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121441 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P679860 AK147960.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P679860 NM_025992.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P679860 AK161293.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P679860 AK147960.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P679860 NM_025992.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P679860 AK161293.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P64514 AK149648.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P64514 AK147960.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P64514 AK011047.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P64514 AK015214.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P64514 AK017888.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P64514 AK149648.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P64514 AK147960.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P64514 AK011047.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P64514 AK015214.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P64514 AK017888.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121441 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109368 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01338034 - -
Whole Mouse Genome G4122A A_52_P679860 AK147960.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P679860 NM_025992.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P679860 AK161293.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P679860 AK147960.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P679860 NM_025992.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P679860 AK161293.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P169908 AK037442.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121441 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109368 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479570 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479564 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479414 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479406 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479352 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479413 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05092222 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479572 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479355 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479344 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479351 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479419 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479357 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05731571 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479560 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479565 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05309411 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479380 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05413805 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479365 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479563 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479348 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479367 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479373 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479368 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479576 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479374 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479418 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479416 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05579596 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479411 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479364 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479562 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479409 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479633 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479377 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479346 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479561 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479354 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479349 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05435617 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479361 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479366 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479567 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479573 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479345 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479360 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24479574 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1431095_PM_a_at - -
Affymetrix HT MG-430 PM Array Plate 1453757_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1432026_PM_a_at - -
Affymetrix HT MG-430 PM Array Plate 1438037_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10538590 - -
Mouse Genome 430 2.0 Array 1453757_PM_at - -
Mouse Genome 430 2.0 Array 1453757_at - -
Mouse Genome 430 2.0 Array 1431095_PM_a_at - -
Mouse Genome 430 2.0 Array 1438037_at - -
Mouse Genome 430 2.0 Array 1438037_PM_at - -
Mouse Genome 430 2.0 Array 1431095_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 67138_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 67138_at - -
Affymetrix Mouse Clariom S Array PSR0600005373.mm.1 TC0600003503.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600005373.mm.1 NM_025992.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600005373.mm.1 NM_025992.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600005374.mm.1 TC0600003503.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600005374.mm.1 NM_025992.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600005374.mm.1 NM_025992.2 Refseq
Mouse Expression Set 430 1453757_PM_at - -
Mouse Expression Set 430 1438037_at - -
Mouse Expression Set 430 1438037_PM_at - -
Mouse Expression Set 430 1453757_at - -
Mouse Expression Set 430 1431095_PM_a_at - -
Mouse Expression Set 430 1431095_a_at - -
Affymetrix HT Mouse Genome 430B Array 1453757_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432026_PM_a_at - -
Affymetrix HT Mouse Genome 430B Array 1453757_at - -
Affymetrix HT Mouse Genome 430B Array 1438037_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432026_a_at - -
Affymetrix HT Mouse Genome 430B Array 1438037_at - -
Affymetrix HT Mouse Genome 430B Array 1431095_PM_a_at - -
Affymetrix HT Mouse Genome 430B Array 1431095_a_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 745835 - -
Array Name Probe IDs Targets Target Type
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2609101 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2609101 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8mgvv:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0