This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01342205_m1
See other IL1RAP GE Assays ›
Gene Symbol:
Il1rap
Gene Name:
interleukin 1 receptor accessory protein
Gene Aliases:
6430709H04Rik, AI255955, AV239853, IL-1RAcP
Chromosome Location:
Chr.16: 26581688 - 26730128 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01342205_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il1rap
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159317.1 NP_001152789.1 2 - 3 250 63
NM_001159318.1 NP_001152790.1 2 - 3 250 63
NM_008364.2 NP_032390.1 2 - 3 250 63
NM_134103.2 NP_598864.1 2 - 3 250 63
GenBank mRNA AK033972.1 - 2 - 3 199 63
AK034269.1 - 2 - 3 214 63
AK039582.1 - 2 - 3 202 63
AK045686.1 - 2 - 3 199 63
AK083209.1 - 2 - 3 247 63
AK136782.1 - 2 - 3 201 63
AK149467.1 - 2 - 3 199 63
BC021159.1 - 2 - 3 199 4161899 63
BU526001.1 - 2 - 3 250 63
X85999.1 - 2 - 3 201 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000376.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Il1rap

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

immune system process
inflammatory response
signal transduction
cytokine-mediated signaling pathway
interleukin-2 biosynthetic process
innate immune response
signal transducer activity
interleukin-1 receptor activity
interleukin-1 receptor binding
protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600003568.mm.1 NM_001159317.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003568.mm.1 NM_001159317.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003568.mm.1 NM_001159318.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003568.mm.1 NM_001159318.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003568.mm.1 NM_008364.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003568.mm.1 NM_008364.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600003568.mm.1 NM_134103.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003568.mm.1 NM_134103.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600003568.mm.1 TC1600000376.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600003567.mm.1 NM_001159317.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003567.mm.1 NM_001159317.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003567.mm.1 NM_001159318.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003567.mm.1 NM_001159318.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003567.mm.1 NM_008364.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003567.mm.1 NM_008364.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600003567.mm.1 NM_134103.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003567.mm.1 NM_134103.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600003567.mm.1 TC1600000376.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600003565.mm.1 NM_001159317.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003565.mm.1 NM_001159317.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003565.mm.1 NM_001159318.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003565.mm.1 NM_001159318.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003565.mm.1 NM_008364.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003565.mm.1 NM_008364.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600003565.mm.1 NM_134103.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003565.mm.1 NM_134103.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600003565.mm.1 TC1600000376.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600003561.mm.1 NM_001159317.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003561.mm.1 NM_001159317.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003561.mm.1 NM_001159318.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003561.mm.1 NM_001159318.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003561.mm.1 NM_008364.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003561.mm.1 NM_008364.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600003561.mm.1 NM_134103.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003561.mm.1 NM_134103.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600003561.mm.1 TC1600000376.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 337702 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0