This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01342342_g1
See other GRK6 GE Assays ›
Gene Symbol:
Grk6
Gene Name:
G protein-coupled receptor kinase 6
Gene Aliases:
Gprk6
Chromosome Location:
Chr.13: 55445072 - 55460927 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01342342_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Grk6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038018.4 NP_001033107.1 10 - 11 1152 74
NM_001112711.2 NP_001106182.1 10 - 11 1152 74
NM_001286063.1 NP_001272992.1 10 - 11 1132 74
NM_001286064.1 NP_001272993.1 10 - 11 1137 74
NM_001286065.1 NP_001272994.1 10 - 11 1050 74
NM_001286066.1 NP_001272995.1 10 - 11 1145 74
NM_011938.4 NP_036068.2 10 - 11 1152 74
GenBank mRNA AF040747.1 - 10 - 11 1022 74
AF040748.1 - 10 - 11 1022 74
AF040749.1 - 10 - 11 1022 74
AK051405.1 - 10 - 11 1134 74
AK146119.1 - 10 - 11 925 74
AK161142.1 - 8 - 9 784 74
AK192356.1 - 2 - 3 117 74
BC057206.1 - 10 - 11 1126 74
BC075719.1 - 10 - 11 1033 5031455 74
Y15797.1 - 10 - 11 974 74
Y15798.1 - 10 - 11 996 74
Y15799.1 - 10 - 11 979 74
Y15800.1 - 10 - 11 967 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000660.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Grk6

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

desensitization of G-protein coupled receptor protein signaling pathway
protein phosphorylation
signal transduction
negative regulation of anion channel activity
Wnt signaling pathway
phosphorylation
termination of G-protein coupled receptor signaling pathway
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
G-protein coupled receptor kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
beta-adrenergic receptor kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300004378.mm.1 NM_001038018.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300004378.mm.1 NM_001112711.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300004378.mm.1 NM_001286063.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004378.mm.1 NM_001286064.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004378.mm.1 NM_001286065.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004378.mm.1 NM_001286066.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004378.mm.1 NM_011938.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300004378.mm.1 TC1300000660.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300004379.mm.1 NM_001038018.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300004379.mm.1 NM_001112711.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300004379.mm.1 NM_001286063.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004379.mm.1 NM_001286064.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004379.mm.1 NM_001286065.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004379.mm.1 NM_001286066.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004379.mm.1 NM_011938.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300004379.mm.1 TC1300000660.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline