Assay Details
| Gene Symbol: Prkdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011159.2 | NP_035289.2 | 67 - 68 | 9469 | 138 | |
| GenBank mRNA | AB007544.1 | - | 67 - 68 | 9446 | 138 | |
| AB011543.1 | - | 67 - 68 | 9458 | 138 | ||
| D87521.1 | - | 67 - 68 | 9446 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkdc
Genomic Map