This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01343110_m1
See other BAAT GE Assays ›
Gene Symbol:
Baat
Gene Name:
bile acid-Coenzyme A: amino acid N-acyltransferase
Gene Aliases:
AI118337, AI158864, BAT
Chromosome Location:
Chr.4: 49489416 - 49507912 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01343110_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Baat
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007519.3 NP_031545.2 3 - 4 956 76
GenBank mRNA AK017923.1 - 3 - 4 906 76
BC012683.1 - 3 - 4 903 76
CB947692.1 - 2 - 3 281 76
U95215.1 - 3 - 4 861 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002636.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Baat

Panther Classification:

Molecular Function -

esterase hydrolase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

bile acid conjugation
glycine metabolic process
lipid metabolic process
fatty acid metabolic process
acyl-CoA metabolic process
bile acid biosynthetic process
bile acid metabolic process
taurine metabolic process
receptor binding
palmitoyl-CoA hydrolase activity
N-acyltransferase activity
transferase activity
transferase activity, transferring acyl groups
hydrolase activity
thiolester hydrolase activity
glycine N-choloyltransferase activity
carboxylic ester hydrolase activity
medium-chain acyl-CoA hydrolase activity
long-chain acyl-CoA hydrolase activity
very long chain acyl-CoA hydrolase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 U95215.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 AK017923.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 BC012683.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 NM_007519.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 AA255212.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 U95215.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 AK017923.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 BC012683.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 NM_007519.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 AA255212.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 716304 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 580401 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 441368 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207612 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 101287 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 512202 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 103082 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647033 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174035 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8049 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130351 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100866 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 U95215.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 AK017923.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 BC012683.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 NM_007519.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 AA255212.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 U95215.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 AK017923.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 BC012683.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 NM_007519.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 AA255212.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P245123 U95215.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905447 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905443 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905433 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905451 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905455 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905469 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905466 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905445 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905438 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905449 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905431 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905440 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905468 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905430 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905452 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905435 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905448 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905429 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905439 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905436 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905441 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05231798 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130351 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100866 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05231820 - -
Whole Mouse Genome G4122A A_51_P245123 U95215.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P43150 U95215.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P43150 AK017923.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P43150 BC012683.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P43150 NM_007519.3 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P43150 AA255212.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P43150 U95215.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P43150 AK017923.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P43150 BC012683.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P43150 NM_007519.3 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P43150 AA255212.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112408 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130351 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05231798 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100866 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04023398 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03323081 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04023399 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04023394 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04023397 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03323084 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905439 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905399 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905344 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905438 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905441 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905430 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905374 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905437 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905489 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905381 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905363 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905480 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905398 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905484 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905346 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905414 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905406 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905409 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905383 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905386 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905403 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905440 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905375 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905431 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905377 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905413 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905339 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905366 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905427 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905481 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905494 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905468 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905421 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905449 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905347 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905401 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905349 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905482 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905373 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905425 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905443 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905380 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905400 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905392 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905387 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905376 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905451 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905405 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905448 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905416 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905379 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905487 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905435 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905469 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905423 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905341 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905447 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905418 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905372 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905428 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905388 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905368 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905445 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905491 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03420639 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905421 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905425 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905413 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03961280 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905406 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905423 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03961289 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905471 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905447 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905368 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03632649 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905343 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905445 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905356 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905435 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905405 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905366 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905369 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905441 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905470 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04023399 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905433 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905427 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04023784 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03314084 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905407 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03246355 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03246363 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905400 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905466 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03420644 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905443 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03590552 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905403 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905398 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905351 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905396 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905438 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905365 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905455 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905448 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905430 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905344 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905418 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905436 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03246348 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905341 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905431 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905363 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905469 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03961281 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905409 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03323083 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905401 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03961283 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905349 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22905468 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10512896 - -
Affymetrix Mouse Gene 1.0 ST Array 10512900 - -
Affymetrix Mouse Gene 1.0 ST Array 10512899 - -
Affymetrix Mouse Gene 1.0 ST Array 10512898 - -
Affymetrix Mouse Gene 1.0 ST Array 10512897 - -
Affymetrix HT MG-430 PM Array Plate 1419001_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1419002_PM_s_at - -
Affymetrix Mouse Gene 1.1 ST Array 10512895 - -
Mouse Genome 430 2.0 Array 1419002_s_at - -
Mouse Genome 430 2.0 Array 1419001_at - -
Mouse Genome 430 2.0 Array 1419001_PM_at - -
Mouse Genome 430 2.0 Array 1419002_PM_s_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 12012_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 12012_PM_at - -
Affymetrix Mouse Clariom S Array PSR0400021468.mm.1 TC0400002636.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400021468.mm.1 NM_007519.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400021468.mm.1 NM_007519.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400021469.mm.1 TC0400002636.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400021469.mm.1 NM_007519.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400021469.mm.1 NM_007519.3 Refseq
Mouse Expression Set 430 1419001_PM_at - -
Mouse Expression Set 430 1419002_s_at - -
Mouse Expression Set 430 1419002_PM_s_at - -
Mouse Expression Set 430 1419001_at - -
Murine Genome U74v2 Set 104273_at - -
Murine Genome U74v2 Set 104273_PM_at - -
Mouse Genome 430A 2.0 Array 1419002_s_at - -
Mouse Genome 430A 2.0 Array 1419001_at - -
Mouse Genome 430A 2.0 Array 1419002_PM_s_at - -
Mouse Genome 430A 2.0 Array 1419001_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1419002_s_at - -
Affymetrix HT Mouse Genome 430A Array 1419001_at - -
Affymetrix HT Mouse Genome 430A Array 1419002_PM_s_at - -
Affymetrix HT Mouse Genome 430A Array 1419001_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 565059 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 730739 - -
Illumina mouse-6 v1.0 expression beadchip 730739 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000730739 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259009 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1259009 - -
mouse-6 v1.1 (Illumina) scl24338.5.1_130-S - -
Illumina Sentrix mouse-6.v1 bead array 730739 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl24338.5.1_130-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 730739 - -
MouseRef-8 v2 Expression BeadChip ILMN_1259009 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1259009 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0