This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01343141_m1
See other EHHADH GE Assays ›
Gene Symbol:
Ehhadh
Gene Name:
enoyl-Coenzyme A, hydratase/3-hydroxyacyl Coenzyme A dehydrogenase
Gene Aliases:
1300002P22Rik, HD, L-PBE, LBFP, LBP, MFP, MFP1, PBFE
Chromosome Location:
Chr.16: 21761285 - 21787834 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01343141_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ehhadh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023737.3 NP_076226.2 6 - 7 958 71
GenBank mRNA AK004867.1 - 6 - 7 929 71
BC016899.1 - 6 - 7 932 4241677 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001368.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ehhadh

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

internal protein amino acid acetylation
lipid metabolic process
fatty acid metabolic process
fatty acid beta-oxidation
acyl-CoA metabolic process
metabolic process
oxidation-reduction process
catalytic activity
3-hydroxyacyl-CoA dehydrogenase activity
dodecenoyl-CoA delta-isomerase activity
enoyl-CoA hydratase activity
receptor binding
oxidoreductase activity
lyase activity
isomerase activity
enzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600011328.mm.1 NM_023737.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600011328.mm.1 NM_023737.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600011328.mm.1 TC1600001368.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0