This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01343163_m1
See other LIPH GE Assays ›
Gene Symbol:
Liph
Gene Name:
lipase, member H
Gene Aliases:
C130037N08Rik, D16Wsu119e, H06, Lpdlr, P3, PLA1B, lpd1, lpd2, mPA-PLA1
Chromosome Location:
Chr.16: 21953817 - 21995542 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01343163_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Liph
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001083894.1 NP_001077363.1 3 - 4 697 85
NM_001289581.1 NP_001276510.1 3 - 4 756 85
NM_001289582.1 NP_001276511.1 3 - 4 697 85
NM_153404.3 NP_700453.1 3 - 4 697 85
GenBank mRNA AK035498.1 - 3 - 4 650 85
AK048150.1 - 3 - 4 596 85
AK134352.1 - 3 - 4 656 85
AK136503.1 - 3 - 4 660 85
AK137278.1 - 3 - 4 728 85
AK161067.1 - 3 - 4 641 85
AK162492.1 - 3 - 4 663 85
AY093499.1 - 3 - 4 524 85
BC037489.1 - 3 - 4 615 4487939 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001374.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Liph

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
lipid catabolic process
phospholipase activity
heparin binding
prenylcysteine methylesterase activity
lipase activity
hydrolase activity
1-oxa-2-oxocycloheptane lactonase activity
sulfolactone hydrolase activity
3,4-dihydrocoumarin hydrolase activity
butyrolactone hydrolase activity
endosulfan lactone lactonase activity
L-ascorbate 6-phosphate lactonase activity
Ser-tRNA(Thr) hydrolase activity
Ala-tRNA(Pro) hydrolase activity
Cys-tRNA(Pro) hydrolase activity
Ser(Gly)-tRNA(Ala) hydrolase activity
all-trans-retinyl-palmitate hydrolase, all-trans-retinol forming activity
retinyl-palmitate esterase activity
carboxylic ester hydrolase activity
mannosyl-oligosaccharide 1,6-alpha-mannosidase activity
mannosyl-oligosaccharide 1,3-alpha-mannosidase activity
methyl indole-3-acetate esterase activity
methyl salicylate esterase activity
methyl jasmonate esterase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600011367.mm.1 NM_001289581.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600011367.mm.1 TC1600001374.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600011365.mm.1 NM_001083894.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600011365.mm.1 NM_001083894.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600011365.mm.1 NM_001289581.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600011365.mm.1 NM_001289582.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600011365.mm.1 NM_153404.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600011365.mm.1 TC1600001374.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600011366.mm.1 NM_001083894.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600011366.mm.1 NM_001083894.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600011366.mm.1 NM_001289581.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600011366.mm.1 NM_001289582.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600011366.mm.1 NM_153404.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600011366.mm.1 TC1600001374.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0