This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01343231_gH
See other PDE4C GE Assays ›
Gene Symbol:
Pde4c
Gene Name:
phosphodiesterase 4C, cAMP specific
Gene Aliases:
Dpde1, E130301F19Rik, dunce
Chromosome Location:
Chr.8: 70723689 - 70751186 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01343231_gH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde4c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_201607.2 NP_963901.1 13 - 14 1501 71
GenBank mRNA AK053713.1 - 7 - 8 2072 71
AK149514.1 - 13 - 14 1451 71
BC030873.1 - 13 - 14 1501 4223352 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000763.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde4c

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

cAMP catabolic process
signal transduction
negative regulation of insulin secretion
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
phosphoric diester hydrolase activity
drug binding
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800005431.mm.1 NM_201607.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800005431.mm.1 NM_201607.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800005431.mm.1 TC0800000763.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline