This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01343473_m1
See other TMPRSS15 GE Assays ›
Gene Symbol:
Tmprss15
Gene Name:
transmembrane protease, serine 15
Gene Aliases:
Entk, Prss7
Chromosome Location:
Chr.16: 78953008 - 79091097 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01343473_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmprss15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008941.3 NP_032967.1 15 - 16 1738 83
NM_178855.4 NP_849186.2 14 - 15 1693 83
GenBank mRNA AK033582.1 - 14 - 15 1695 83
AK038356.1 - 6 - 7 687 83
BC117917.1 - 15 - 16 1737 40097367 83
BC117918.1 - 15 - 16 1737 40097370 83
U73378.1 - 15 - 16 1733 83

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmprss15

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity
scavenger receptor activity
peptidase activity
serine-type peptidase activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0