This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01343781_m1
See other SIDT1 GE Assays ›
Gene Symbol:
Sidt1
Gene Name:
SID1 transmembrane family, member 1
Gene Aliases:
AW045928, B830021E24Rik
Chromosome Location:
Chr.16: 44240177 - 44333328 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01343781_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sidt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159419.1 NP_001152891.1 22 - 23 2416 64
NM_198034.3 NP_932151.2 21 - 22 2401 64
GenBank mRNA AK034344.1 - 7 - 8 566 64
AK171920.1 - 21 - 22 2554 64
AY562213.1 - 11 - 12 960 64
AY562214.1 - 11 - 12 960 64
BC079584.1 - 21 - 22 2401 5704672 64

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sidt1

Gene Ontology Categories:

Function(s) Process(es)

biological_process
dsRNA transport
molecular_function
RNA transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline