This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01343865_m1
See other CD200 GE Assays ›
Gene Symbol:
Cd200
Gene Name:
CD200 antigen
Gene Aliases:
Mox2, OX2
Chromosome Location:
Chr.16: 45382111 - 45409058 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01343865_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd200
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010818.3 NP_034948.3 1 - 2 195 62
GenBank mRNA AF004023.1 - 1 - 2 33 62
AK148308.1 - 1 - 2 164 62
AK164478.1 - 1 - 2 118 62
BC019172.1 - 1 - 2 65 5003655 62
BC051984.1 - 1 - 2 113 5707683 62
BC054759.1 - 1 - 2 128 6413363 62
CN665640.1 - 1 - 2 195 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF004023.1, AK148308.1, AK164478.1, BC019172.1, BC051984.1, BC054759.1, NM_010818.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cd200

Gene Ontology Categories:

Process(es)

protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600013550.mm.1 NM_010818.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600013550.mm.1 NM_010818.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600013550.mm.1 TC1600001655.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600013541.mm.1 NM_010818.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600013541.mm.1 NM_010818.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600013541.mm.1 TC1600001655.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600013549.mm.1 NM_010818.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600013549.mm.1 NM_010818.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600013549.mm.1 TC1600001655.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 744720 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline