This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01343879_m1
See other CD200R1 GE Assays ›
Gene Symbol:
Cd200r1
Gene Name:
CD200 receptor 1
Gene Aliases:
CD200R, Mox2r, OX2R
Chromosome Location:
Chr.16: 44765736 - 44794977 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01343879_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd200r1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021325.3 NP_067300.1 4 - 5 969 149
GenBank mRNA AK089173.1 - 4 - 5 906 149
AK154325.1 - 4 - 5 960 149
AK154722.1 - 4 - 5 928 149
AK171130.1 - 4 - 5 901 149
AK171590.1 - 4 - 5 905 149
BC052682.1 - 4 - 5 927 30072503 149

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000662.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cd200r1

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Process(es)

receptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600005809.mm.1 NM_021325.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005809.mm.1 NM_021325.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600005809.mm.1 TC1600000662.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600005812.mm.1 NM_021325.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005812.mm.1 NM_021325.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600005812.mm.1 TC1600000662.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline